Learn More
Cayman Chemical EthIdIumbromIde 1g
SDP
Supplier:  Cayman Chemical 391321 G
Encompass_Preferred
A fluorescent dye and DNA intercalator; ex/em = 525/595 nm, respectively; inhibits the DNA-unwinding and ATPase activities of P. falciparum DNA helicase (IC50s = 1.5 and 2.5 µM, respectively) and is active against P. falciparum in vitro; completely inhibits E. coli RNA polymerase at 100 µmol/ml; active against Mycoplasma from 0.5-8 UG/ml; increases lifespan in 6C3HED-OG and L5178Y murine lymphoma models at 8 mg/kg; has commonly been used to stain DNA in agarose gel electrophoresis and in displacement assays to determine compound binding to DNA
Catalog No. 50-291-8553