Learn More
Medchemexpress LLC Ethionamide-d3 | 2748267-66-3 | >=99% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0276S1MG
Encompass_Preferred
Ethionamide-d3 is the deuterium labeled Ethionamide, an antibiotic utilized in the treatment of tuberculosis. This compound is intended for research use only and can be employed as a tracer or an internal standard for quantitative analysis. Deuteration, the incorporation of stable heavy isotopes, has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Active against *M. tuberculosis*.
  • Not for human or veterinary use.
Catalog No. 50-004-06458