Learn More
Medchemexpress LLC Ethiprole | 181587-01-9 | 95.96% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12042550MG
Encompass_Preferred
Ethiprole is an analytical/reference standard of the insecticide Ethiprole (CAS 181587-01-9) supplied as a solid for use in qualitative and quantitative analytical methods, including LC, GC, and MS. It has formula C13H9Cl2F3N4OS and molecular weight 397.20.
  • Analytical/reference standard suitable for method development and calibration.
  • High purity for reliable analytical results (>98.0%).
  • Available in small pack sizes for laboratory use (50 mg and larger).
  • White to light yellow solid appearance simplifies handling and identification.
  • Applicable to environmental residue analysis and pesticide monitoring.
Catalog No. 50-002-33345