Learn More
Medchemexpress LLC Ethiprole | 181587-01-9 | ≥98.0% | 100 MG

Supplier: Medchemexpress LLC HY120425100MG
Ethiprole is an analytical reference standard of the insecticide Ethiprole intended for laboratory analytical testing and method development. It is supplied as a small mass package (100 mg) and is provided as an analytical/reference standard with assay information available in the Certificate of Analysis.
- Analytical reference standard for analytical testing and method development.
- CAS number 181587-01-9.
- Chemical formula C13H9Cl2F3N4OS; molecular weight 397.20 g/mol.
- Available in small mass packages, for example 100 mg.
- Certificate of Analysis provides assay and recommended storage conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.