Learn More
Cayman Chemical Ethoprop-d6 10mg
SDP
Supplier:  Cayman Chemical 3960110 MG
Encompass_Preferred
An internal standard for the quantification of ethoprop by GC- or LC-MS
Catalog No. 50-291-7167