Learn More
Medchemexpress LLC Ethopropazine hydrochloride | 1094-08-2 | MFCD00012653 | 99.3% | 348.93 g/mol | C19H25ClN2S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10792225MG
Encompass_Preferred
Ethopropazine hydrochloride is the hydrochloride salt of ethopropazine, a phenothiazine-derived research reagent used as a butyrylcholinesterase (BChE) inhibitor and a non-selective muscarinic acetylcholine (mAChR) and NMDA receptor antagonist. It is supplied as a white to off-white solid (CAS 1094-08-2), molecular formula C19H25ClN2S, molecular weight 348.93 g/mol, and is offered in solid and DMSO solution forms with manufacturer-listed purity ~99.3%.
  • High purity suitable for biochemical and pharmacological studies.
  • Available as solid and as 10 mM solutions in DMSO.
  • Stable when stored sealed at 4°C; in solvent store at -80°C for long term.
  • Useful as a BChE inhibitor and for cholinergic and NMDA pathway assays.
  • White to off-white solid, easy to handle for laboratory use.
Catalog No. 50-002-42288