Learn More
Cayman Chemical EthosuxImIde 500mg
SDP
Supplier:  Cayman Chemical 23947500 MG
Encompass_Preferred
An anticonvulsant; increases glucose, fructose-1,6-bisphosphate, and pyruvate levels in rat brain at 200 mg/kg; reduces the severity of audiogenic seizures in a rat model of barbiturate withdrawal-induced convulsions at 400 mg/kg; inhibits PTZ- or brainstem stimulation-induced tonic hindlimb extension (ED50s = 35 and 70 mg/kg, respectively), as well as leptazol-induced clonic seizures (ED50 = 230 mg/kg), in rats; reduces resting tremor by 60% in a macaque model of Parkinson's disease induced by MPTP when administered at a dose of 150 mg/animal
Catalog No. 50-296-4420