Learn More
Medchemexpress LLC Ethyl 1-[(4-acetamidophenyl)methyl]-4-(2-phenylethyl)piperidine-4-carboxylate | 1069498-96-9 | 99.1% | 408.53 g/mol | C25H32N2O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10400625MG
Encompass_Preferred
CYM51010 is a research compound that acts as a biased ligand for μ-opioid receptor-δ-opioid receptor heterodimers. It demonstrates anti-nociceptive activity similar to morphine but with reduced tolerance development and withdrawal symptoms, and is supplied as a high-purity solid for laboratory use.
  • Biased μ-δ opioid receptor ligand for studying heterodimer signaling.
  • Exhibits anti-nociceptive activity with reduced tolerance and withdrawal.
  • High reported purity suitable for experimental applications.
  • Soluble in DMSO at high concentration for convenient stock preparation.
  • Supplied as a solid with cold, light-protected storage recommendations.
Catalog No. 50-002-44477