Learn More
eMolecules​ Ethyl 2-(1,3-dioxoisoindol-2-yl)acetate | Combi-Blocks | 6974-10-3 | MFCD00023083 | 233.223 | C12H11NO4 | 98.000 | CCOC(=O)CN1C(=O)c2ccccc2C1=O | 5g | 335353451
Supplier:  eMolecules​ 335353451
Encompass_Preferred
Ethyl 2-(1,3-dioxoisoindol-2-yl)acetate | Combi-Blocks | 6974-10-3 | MFCD00023083 | 233.223 | C12H11NO4 | 98.000 | CCOC(=O)CN1C(=O)c2ccccc2C1=O | 5g | 335353451
Catalog No. NC1643988