Learn More
Medchemexpress LLC Ethyl (2-amino-3-fluoro-4-((4-(trifluoromethyl)benzyl)amino)phenyl)carbamate | 1919050-87-5 | 99.2% | 371.33 g/mol | C17H17F4N3O2 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12326410MM 1ML
Encompass_Preferred
RL648_81 is a small-molecule activator of KCNQ2/3 potassium channels supplied as a 10 mM solution in DMSO for in vitro research. It shifts the V1/2 of KCNQ2/3 channels toward hyperpolarized potentials and shows an EC50 of about 190 nM. Molecular weight is 371.33 g·mol⁻1, chemical formula C17H17F4N3O2, and analytical purity 99.2%.
Catalog No. 50-002-39530