Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 2-amino-3-hydroxybutanoate hydrochloride | 39994-70-2 | MFCD07437964 | >97.0% | 183.63 g/mol | C6H14ClNO3 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW04033310G

Encompass_Preferred

(2S,3R)-Ethyl 2-amino-3-hydroxybutanoate hydrochloride is the ethyl ester hydrochloride derivative of threonine, provided as a research-grade building block and intermediate for organic synthesis and biochemical studies.

  • Purity ≥97.0%.
  • Molecular weight 183.63 g/mol.
  • Chemical formula C6H14ClNO3.
  • Appearance viscous liquid, colorless to light yellow.
  • Suitable as a synthetic intermediate and building block for peptide and small-molecule synthesis.
  • For research use only; not for human or therapeutic use.

Catalog No. 50-002-08778


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.