Learn More
Medchemexpress LLC Ethyl 2-chloropyrimidine-5-carboxylate | 89793-12-4 | 186.60 | 5 G

Supplier: Medchemexpress LLC HYW0039955G
Ethyl 2-chloropyrimidine-5-carboxylate is a chemical reagent primarily used for synthesis. It is supplied as a solid, appearing white to yellow in color.
- Purity: 99.71% or 99.78%
- Molecular weight: 186.60
- Formula: C7H7ClN2O2
- Appearance: Solid
- Color: White to yellow
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.