Learn More
Medchemexpress LLC Ethyl 2-methylnicotinate | 1721-26-2 | 99.4% | 165.19 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01010510G
Encompass_Preferred
Ethyl 2-methylnicotinate is a biochemical reagent that can be utilized as a biological material or organic compound for life science related research. It is intended for research use only and not sold to patients. Ethyl 2-methylpyridine-3-carboxylate, a related compound, has been employed in the synthesis of amino esters like ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, and ethyl 2-(diethylaminomethyl)benzoate.
  • Utilized as a biological material or organic compound for life science related research.
  • Intended for research use only.
  • Can be employed in the synthesis of amino esters.
Catalog No. 50-004-52478