Learn More
Medchemexpress LLC Ethyl 3,4,5-trimethoxybenzoate | 6178-44-5 | 99.7% | 240.26 | 1 ML

Supplier: Medchemexpress LLC HYW01209210MM/1ML
Ethyl 3,4,5-trimethoxybenzoate is a natural compound isolated from the roots of Rauvolfia yunnanensis Tsiang. It is intended for research use only.
- Natural compound
- Isolated from Rauvolfia yunnanensis Tsiang roots
- For research use only
- High purity (99.67%)
- Molecular weight of 240.26
- Appears as a white to off-white solid
- Soluble in DMSO (≥ 100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.