Learn More
Medchemexpress LLC Ethyl 3-coumarincarboxylate | 1846-76-0 | MFCD00016964 | 99.8% | 218.21 g/mol | C12H10O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01408110MM/1ML
Encompass_Preferred
Ethyl 3-coumarincarboxylate is a coumarin-derived reagent used in organic synthesis and analytical chemistry. It is commonly employed as a pseudo-template for preparing molecularly imprinted polymers (MIPs) with selective recognition for aflatoxins. Available in solid form and as concentrated DMSO solutions, it is supplied with characterization data suitable for research applications.
  • Coumarin derivative useful in synthetic chemistry and method development.
  • Functions as a pseudo-template for molecularly imprinted polymer production.
  • Available as solid and as concentrated solution for convenient handling.
  • High purity suitable for research applications.
  • Well characterized chemical properties (C12H10O4, 218.21 g/mol).
Catalog No. 50-003-03817