Learn More
Medchemexpress LLC Ethyl 3-hydroxybutyrate | 5405-41-4 | 100.0% | 132.16 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01270110G
Encompass_Preferred
Ethyl 3-hydroxybutyrate is a fragrance found in wine and Tribolium castaneum. It has shown potential in alleviating skeletal muscle wasting in cancer cachexia by increasing 3-HB levels in serum, promoting the TCA cycle and protein synthesis, improving metabolic homeostasis, reducing inflammation, and enhancing antioxidant capacity.
  • Increases 3-HB levels in serum.
  • Promotes TCA cycle and protein synthesis.
  • Improves metabolic homeostasis.
  • Reduces inflammation responses.
  • Enhances antioxidant capacity.
  • Alleviates cancer cachexia-associated skeletal muscle atrophy.
Catalog No. 50-004-54920