Learn More
Medchemexpress LLC Ethyl 3-hydroxybutyrate | 5405-41-4 | 100.0% | 132.16 | 25 G

Supplier: Medchemexpress LLC 25GHYW012701
Ethyl 3-hydroxybutyrate is a fragrance found in wine and *Tribolium castaneum*. It is intended for research use only.
- Molecular weight: 132.16
- Purity: 99.99%
- Increases 3-HB levels in serum and promotes TCA cycle and protein synthesis in CT26 xenograft mouse models.
- Improves metabolic homeostasis, reduces inflammation responses, and enhances antioxidant capacity.
- Alleviates cancer cachexia-associated skeletal muscle atrophy in CT26 xenograft mouse models.
- Available in various quantities and solution formats.
- Can be stored at -20°C for 2 years (pure form) or 1 month (in solvent), and at -80°C for 6 months (in solvent).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.