Learn More
Medchemexpress LLC Ethyl 3-oxopentanoate | 4949-44-4 | 97.9% | C7H12O3 | 5 G
SDP
Supplier:  Medchemexpress LLC HY782115G
Encompass_Preferred
Ethyl 3-oxopentanoate is an organic ester compound commonly used as a reactant or solvent in organic synthesis. It serves as an important raw material for the preparation of pharmaceuticals, food flavors, spices, and other products. It is also widely used in some industrial productions due to its stability and reactivity in certain chemical reactions. It is for research use only.
  • Used as a reactant or solvent in organic synthesis
  • Important raw material for pharmaceuticals
  • Important raw material for food flavors and spices
  • Widely used in industrial productions
  • Plays a role in chemical research and industry
Catalog No. 50-004-34880