Learn More
Medchemexpress LLC Ethyl 4-(3-(4-(1H-pyrazol-1-yl)benzyl)ureido)benzoate | 1005883-72-6 | MFCD04280228 | 98.1% | 364.40 g/mol | C20H20N4O3 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12607410MM 1ML
Encompass_Preferred
Z433927330 is a small-molecule inhibitor selective for aquaporin-7 (AQP7), with reduced activity against AQP3 and AQP9. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for in vitro research. Key chemical attributes include molecular formula C20H20N4O3, molecular weight 364.40 g/mol, and reported purity of about 98.1%. Reported IC50 values show strong activity against mAQP7 and lower potency for mAQP3 and mAQP9.
Catalog No. 50-002-34963