Learn More
Medchemexpress LLC Ethyl 4-(butylamino)benzoate | 94-32-6 | MFCD00017283 | 98% | 221.29 | 5 G

Supplier: Medchemexpress LLC HY414625G
Ethyl 4-(butylamino)benzoate is an ethyl ester of 4-(butylamino)-benzoic acid that is utilized in chemical synthesis. It is a chemical compound with the molecular formula C13H19NO2 and appears as a light yellow powder. It may cause irritation and allergic reactions in sensitive individuals.
- Assay: 98%
- Form: Powder
- Boiling point: 220 °C/2 mmHg (lit.)
- Melting point: 68-70 °C (lit.)
- Functional group: Amine, ester
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.