Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl (4-hydroxyphenyl)acetate | 17138-28-2 | MFCD00016491 | 99.3% | 180.20 g/mol | C10H12O3 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW01648310G

Encompass_Preferred

Ethyl (4-hydroxyphenyl)acetate (CAS 17138-28-2) is an organic reagent used as a carbon source and a synthetic building block in biochemical and microbiological research. It is colorless to light yellow, solid below ~36°C and liquid above ~38°C, with a molecular weight of 180.20 g/mol and high reported purity suitable for research applications.

  • Molecular weight 180.20 g/mol.
  • Chemical formula C10H12O3.
  • High purity (99.28%).
  • Solid below ~36°C and liquid above ~38°C.
  • Available in 10 g, 25 g, 50 g, and 100 g pack sizes.
  • Suitable as a carbon source and synthetic intermediate for research.

Catalog No. 50-003-03770


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.