Learn More
Medchemexpress LLC Ethyl 4-isocyanatobenzoate-d4 | 98.0% | 195.21 g·mol⁻1 | C10H5D4NO3 | 25 MG

Supplier: Medchemexpress LLC HYW275290S25MG
Ethyl 4-isocyanatobenzoate-d4 is a deuterium-labeled reagent supplied as a fine white to off-white solid for use as an isotopic standard or intermediate in synthetic and analytical research. The labeled compound supports isotope tracing and serves as a mass-shifted standard in quantitative analyses.
- Chemical formula: C10H5D4NO3.
- Molecular weight: 195.21 g·mol⁻1.
- Isotopic labeling: d4 with 99.7% isotopic enrichment.
- Purity: 98.0% (GC).
- Appearance: white to off-white solid.
- Storage: powder -20 °C (up to 3 years); in solvent -80 °C (6 months) or -20 °C (1 month).
- Typical pack size: 25 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.