Learn More
Medchemexpress LLC Ethyl 6-(4-Aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate | 503615-07-4 | 98.6% | C22H22N4O4 | 250 MG

Supplier: Medchemexpress LLC 250MGHY75678
This chemical compound, with CAS number 503615-07-4, has the molecular formula C22H22N4O4 and a molecular weight of 406.43. It exhibits a high purity of 98.61% and appears as a light green to green solid. The compound is identified as 1H-Pyrazolo[3,4-c]pyridine-3-carboxylic acid, 6-(4-aminophenyl)-4,5,6,7-tetrahydro-1-(4-methoxyphenyl)-7-oxo-, ethyl ester.
- CAS number: 503615-07-4
- Molecular formula: C22H22N4O4
- Molecular weight: 406.43
- Purity: 98.61% (LCMS)
- Appearance: light green to green solid
- Storage for solid at 4°C, protected from light
- Storage for solutions at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.