Learn More
Medchemexpress LLC Ethyl 6-bromohexanoate | 25542-62-5 | MFCD00000270 | 99.6% | 223.11 g/mol | C8H15BrO2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW04197050G
Encompass_Preferred
Ethyl 6-bromohexanoate is an alkyl bromide ester used as a linker and building block in organic synthesis, including preparation of proteolysis targeting chimeras (PROTACs) and other small-molecule conjugates. It is supplied for research use and should be handled under standard laboratory safety procedures.
  • High purity suitable for research applications.
  • Commonly used as an alkylating linker in syntheses.
  • Available in multiple pack sizes for scale flexibility.
  • Characterized by CAS 25542-62-5 and formula C8H15BrO2.
Catalog No. 50-002-09559