Learn More
Medchemexpress LLC Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate | 28705-46-6 | MFCD00227484 | 99.8% | 289.33 g/mol | C16H19NO4 | 5 G

Supplier: Medchemexpress LLC HYW0874335G
Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate is a coumarin-derived small molecule reported as a selective inhibitor of human monoamine oxidase B (hMAO-B) with an IC50 of 45.52 μM. It is provided as a high-purity solid for research use and is supplied in multiple pack sizes.
- High purity (99.8%).
- Molecular weight 289.33 g/mol.
- Chemical formula C16H19NO4.
- Reported activity: hMAO-B inhibitor (IC50 = 45.52 μM).
- Available as a solid in a 5 G pack and other sizes.
- Certificate of analysis available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.