Learn More
Medchemexpress LLC Ethyl arachidonate | 1808-26-0 | 98.3% | 332.52 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW01321410MG
Encompass_Preferred
Ethyl arachidonate is a lipophilic esterified form of arachidonic acid (AA). It can be used in dietary regimens or fed to cultured cells as a source of exogenous arachidonate. It is also the main species of fatty acid ethyl esters (FAEE) found in the brain of alcohol-intoxicated subjects.
  • Lipophilic esterified form of arachidonic acid
  • Can be incorporated into dietary regimens
  • Can be fed to cultured cells as a source of exogenous arachidonate
  • Primary fatty acid ethyl ester (FAEE) found in the brains of alcohol-intoxicated subjects
  • Research use only
Catalog No. 50-004-54447