Learn More
Medchemexpress LLC Ethyl Caffeate | 102-37-4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN6966
Encompass_Preferred
Ethyl Caffeate is a natural phenolic compound isolated from Bidens pilosa. It suppresses NF-κB activation and its downstream inflammatory mediators, inducible nitric oxide synthase (iNOS), cyclooxygenase-2 (COX-2), and prostaglandin E2 (PGE2) in vitro or in mouse skin. It is for research use only and not sold to patients.
  • Suppresses NF-κB activation
  • Inhibits inflammatory mediators, including iNOS, COX-2, and PGE2
  • Markedly suppresses LPS-induced nitric oxide (NO) production
  • Targets iNOS, NF-κB, and COX-2
  • Isolated from Bidens pilosa
Catalog No. 50-004-21822