Learn More
Medchemexpress LLC Ethyl cinnamate | 103-36-6 | MFCD00009189 | 176.21 g/mol | C11H12O2 | 500g
SDP
Supplier:  Medchemexpress LLC HYY0121500G
Encompass_Preferred
Ethyl cinnamate an orally active chemical constituent of the rhizome of Kaempferia galanga exhibits anti-cancer nematocidal sedative and vasorelaxant activities Ethyl cinnamate is a fragrance ingredient used as a food flavor and additive for cosmetic products Ethyl cinnamate is also an excellent clearing reagent for mammalian tissues Ethyl cinnamate suppresses tumor growth through anti-angiogenesis by attenuating VEGFR2 signal pathway in colorectal cancer Ethyl cinnamate inhibits the tonic contractions induced by high K and phenylephrine (PE) with respective IC50 values of 0 30 mM and 0 38 mM in rat aorta[1][2][3][4]
Catalog No. 50-002-68187