Learn More
Medchemexpress LLC Ethyl β-D-glucopyranoside | 3198-49-0 | MFCD09838462 | 99.8% | 208.21 | C8H16O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN387210MG
Encompass_Preferred
Ethyl β-D-glucopyranoside is a high-purity glycoside used as a reference standard and reagent in glycobiology and natural product research. It is typically supplied as a white to almost white powder and is characterized by CAS 3198-49-0, molecular formula C8H16O6, and molecular weight 208.21.
  • High purity suitable for analytical applications (≈99.8%).
  • Available in small, lab-scale amounts for research (including 10 MG packs).
  • Used as a reference standard in glycobiology and natural product studies.
  • White to almost white powder with melting point 80-84 °C.
  • Identity confirmed by NMR and GC-based purity analysis.
Catalog No. 50-002-89501