Learn More
Medchemexpress LLC Ethyl diethoxyacetate | 6065-82-3 | 176.21 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW001603
Encompass_Preferred
Ethyl diethoxyacetate is an organic synthesis reagent that shows moderate antiproliferative effects on cervical cancer and liver cancer. It is a liquid with a relative density of 0.985 g/cm3, stable under recommended storage conditions, and is for research use only.
  • Used as an organic synthesis reagent
  • Exhibits moderate antiproliferative effects on cervical cancer and liver cancer
  • Liquid appearance
  • Stable under recommended storage conditions
  • Suitable for laboratory chemical applications
Catalog No. 50-004-49895