Learn More
Medchemexpress LLC Ethyl docos-13-enoat 10g
SDP
Supplier:  Medchemexpress LLC HYW09964410G
Encompass_Preferred
Ethyl docos-13-enoate is a colorless or pale yellow liquid with a sweet floral aroma used as a flavoring agent in the food industry and as a flavor enhancer in perfumes and colognes It is also found in some vegetable oils and is used as a surfactant and emulsifier in personal care products Ethyl docos-13-enoate is a versatile compound used in various industries due to its pleasant odor and ability to mix ingredients together
Catalog No. 50-002-06429