Learn More
Medchemexpress LLC Ethyl (E)-2-hexenoate | 27829-72-7 | MFCD00036541 | 98.5% | 142.20 g/mol | C8H14O2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW017567
Encompass_Preferred
Ethyl (E)-2-hexenoate (CAS 27829-72-7) is the ethyl ester of trans-2-hexenoic acid. It is a colorless liquid used as a flavor and fragrance ingredient, a synthetic building block, and a reagent in organic synthesis and research. Typical supplier-listed purity is about 98.5%.
  • Colorless liquid at room temperature.
  • Molecular formula C8H14O2; molecular weight 142.20 g/mol.
  • High purity suitable for analytical and synthetic use.
  • Commonly used in flavor and fragrance formulation and as a synthetic intermediate.
  • Store under recommended conditions; protect from light and moisture.
Catalog No. 50-003-03829