Learn More
Medchemexpress LLC Ethyl (E)-3-(1-pyrrolidinyl)crotonate | 54716-02-8 | 99.7% | 183.25 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY44087
Encompass_Preferred
Ethyl (E)-3-(1-pyrrolidinyl)crotonate is a biochemical reagent useful as a biological material or organic compound for life science-related research. It has a molecular weight of 183.25 and a formula of C10H17NO2. The product appears as a solid below 25°C and a liquid above 26°C, with a light yellow to light brown color. It is intended for research use only and is not for sale to patients.
  • Biochemical reagent for life science research.
  • Can be used as a biological material.
  • Can be used as an organic compound.
  • High purity: 99.73%.
  • Available in various quantities, including solution and solid forms.
  • Detailed documentation available (Data sheet, COA, SDS, handling instructions).
  • Soluble in DMSO (≥ 200 mg/mL).
Catalog No. 50-004-02188