Learn More
Selleck Chemical LLC Ethyl gallate
SDP
Supplier:  Selleck Chemical LLC S555025MG
Encompass_Preferred
Ethyl gallat (Phyllemblin gallic acid ethyl ester) which could be found naturally in a variety of plant sources is a food additive with antimicrobial activity Ethyl gallat activates the death receptor-dependent pathway of apoptosis by enhancing the expression of caspases-8 -9 and -3 and the Bcl-2 interacting domain (Bid)
Catalog No. 50-000-03110