Learn More
Medchemexpress LLC Ethyl gallate | 831-61-8 | 198.17 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN052510MM/1ML
Encompass_Preferred
Ethyl gallate is a nonflavonoid phenolic compound and a scavenger of hydrogen peroxide. It has a cytotoxic effect on HL-60 cells, leading to changes in morphology, shrinkage of the cell membrane, development of apoptotic bodies, and decreased cell viability. It is suitable for laboratory research applications.
  • Nonflavonoid phenolic compound
  • Scavenger of hydrogen peroxide
  • Has cytotoxic effect on HL-60 cells
  • Leads to changes in morphology, shrinkage of the cell membrane, development of apoptotic bodies, and decreased cell viability
Catalog No. 50-003-90015