Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Glycidyl Ether, Spectrum™ Chemical
SDP

Synonym: 2-(Ethoxymethyl)oxirane, (Ethoxymethyl)oxirane

$124.00 - $370.00

Chemical Identifiers

CAS 4016-11-9
Molecular Weight (g/mol) 102.13 g/mol
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-620-700
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
E146625ML
25 mL
Each for $124.00
Add to Cart
 
18-620-701
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
E1466100ML
100 mL
Each for $370.00
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

4016-11-9
102.13 g/mol

Specifications

4016-11-9
98%
Ethyl Glycidyl Ether
102.13 g/mol
Amber Glass Bottle
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.