Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Glycidyl Ether, Spectrum™ Chemical
SDP

Synonym: 2-(Ethoxymethyl)oxirane, (Ethoxymethyl)oxirane

Supplier:  Spectrum Chemical Manufacturing Corporation E146625ML

Quantity 25 mL

 View more versions of this product

Catalog No. 18-620-700


Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart

Description

Description

Specifications

Chemical Identifiers

4016-11-9
102.13 g/mol

Specifications

4016-11-9
98%
25 mL
102.13 g/mol
Amber Glass Bottle
Ethyl Glycidyl Ether
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $500 in Chemicals with Purchase
Spend $250 or more on Spectrum Chemical chemicals and get up to $500 to spend on additional chemicals.

Adding...

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.