Learn More
Accela Chembio Inc MF: C4H8O3 | MW: 104.10 | ?98% | Flash Point: 62¿C(143.6¿F) | BP: 158-159¿C | Density: 1.1g/mLat25¿C
Supplier:  Accela Chembio Inc SY002762100G
Encompass_Preferred
MF: C4H8O3 | MW: 104.10 | ?98% | Flash Point: 62¿C(143.6¿F) | BP: 158-159¿C | Density: 1.1g/mLat25¿C
Catalog No. 50-353-510