Learn More
Medchemexpress LLC Ethyl linolenate | 1191-41-9 | 306.48 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYN2073500MG
Encompass_Preferred
Ethyl linolenate is a fatty acid ethyl ester (FAEE) that inhibits the cellular production of melanin with an IC50 of 70 μM, demonstrating anti-melanogenesis effects. It significantly blocks Forskolin-induced melanogenesis and inhibits tyrosinase activity. This product is for research use only.
  • Fatty acid ethyl ester (FAEE)
  • Inhibits cellular production of melanin with an IC50 of 70 μM
  • Anti-melanogenesis effects
  • Significantly blocks Forskolin-induced melanogenesis
  • Inhibits tyrosinase activity
  • Purity: 98.0%
Catalog No. 50-004-15235