Learn More
Medchemexpress LLC Ethyl N-(5-benzimidazolyl)carbamate | 6306-71-4 | MFCD00552813 | 99.5% | 205.21 g/mol | C10H11N3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB131310MG
Encompass_Preferred
Lobendazole is a benzimidazole-class anthelmintic supplied for research use. It inhibits Giardia intestinalis strains and is provided as a white to off-white solid with high chemical purity.
  • Anthelmintic active against Giardia intestinalis strains.
  • High purity (99.49%).
  • Molecular weight 205.21 g/mol.
  • Packaged as a compact 10 MG dose for laboratory use.
  • White to off-white solid appearance.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-97979