Learn More
Medchemexpress LLC Ethyl nonanoate | 123-29-5 | 98.0% | 1 ML

Supplier: Medchemexpress LLC HY12962310MM/1ML
Ethyl nonanoate is a rich ester found in spirits, contributing to the pleasant fruity bouquet of alcoholic beverages. It is also an endogenous metabolite.
- Molecular weight: 186.29
- Formula: C11H22O2
- Appearance: Liquid
- Color: Colorless to light yellow
- Initial source: Plants other families, microorganisms, endogenous metabolite
- Storage (pure form): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
- Solubility (in vitro): 100 mg/mL in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.