Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl octanoate-d15 | 1219798-38-5 | 98.5% | 187.36 g/mol | C10H5D15O2 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYW015305S

Encompass_Preferred

Ethyl octanoate-d15 is the deuterium-labeled analogue of ethyl octanoate, supplied as a neat, colorless to light yellow liquid. It is used as an isotopic/internal standard in analytical workflows such as mass spectrometry, metabolic tracing, and quantitative assays, and is provided with batch-specific documentation.

  • Chemical formula C10H5D15O2.
  • Molecular weight 187.36 g/mol.
  • Purity 98.5% as reported on the COA.
  • Suitable as an internal standard for mass spectrometry.
  • Supplied as a neat liquid; density 0.867 g/cm3.
  • Storage stability under recommended conditions; refer to COA and datasheet.

Catalog No. 50-003-02936


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.