Learn More
Cayman Chemical Ethyl OrselInate 1g
SDP
Supplier:  Cayman Chemical 227091 G
Encompass_Preferred
A lichen metabolite and a derivative of lecanoric acid; inhibits the proliferation of human Hep-2 larynx carcinoma, MCF-7 breast cancer, and 786-0 kidney carcinoma cells, as well as B16-F10 murine melanoma and non-cancerous Vero cells (IC50s = 31.2, 70.3, 47.5, 64.8, and 28.1 UG/ml, respectively); toxic to brine shrimp (LC50 = 495 µM)
Catalog No. 50-299-4118