Learn More
Medchemexpress LLC Ethyl Orsellinate | 2524-37-0 | 196.20 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00042710MM/1ML
Encompass_Preferred
Ethyl orsellinate is a lichen metabolite and a derivative of lecanoric acid with antiproliferative and antitumour activities.
  • Bacterial inhibitor
  • Shows cytotoxic activity against A. salina with an LC50 of 495 μM
  • Inhibits Hep-2 cells, MCF-7 breast cancer cells, and 786-0 kidney carcinoma cells
  • Active against B16-F10 murine melanoma and non-cancerous Vero cells
Catalog No. 50-004-47678