Learn More
Medchemexpress LLC Ethyl palmitate | 628-97-7 | MFCD00008996 | 284.48 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYN2086
Encompass_Preferred
Ethyl palmitate, also known as ethyl hexadecanoate, is a fatty acid ethyl ester that functions as a CHIKV virus inhibitor. It has demonstrated anti-inflammatory activity by reducing levels of TNF-α, IL-6, and NF-κB in endotoxemic rats. This product is intended for research use only.
  • Inhibits CHIKV virus.
  • Demonstrates anti-inflammatory activity.
  • Reduces levels of TNF-α, IL-6, and NF-κB.
  • Suitable for research applications.
Catalog No. 50-003-89419