Learn More
Medchemexpress LLC Ethyl trans-2-decenoate | 7367-88-6 | MFCD00015299 | 97.0% | 198.30 g/mol | C12H22O2 | 25 G

Supplier: Medchemexpress LLC HYW12757325G
Ethyl trans-2-decenoate is a trans-2-decenoate ester used as a fragrance and flavoring reagent and as a biochemical reagent for research and synthesis. It is a colorless to pale yellow liquid with a fruity, floral aroma and is supplied at high purity for analytical and preparative applications.
- Purity: 97.0%
- Cas number: 7367-88-6
- Molecular formula: C12H22O2
- Molecular weight: 198.30 g/mol
- Appearance: colorless to pale yellow liquid
- Common uses: flavoring and fragrance ingredient, organic synthesis, and biochemical assays
- Available pack sizes: 5 G, 10 G, 25 G, 50 G
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.