Learn More
Medchemexpress LLC Ethyl trans-4-decenoate | 76649-16-6 | MFCD00015574 | >98.0% | 198.30 g/mol | C12H22O2 | 5 G

Supplier: Medchemexpress LLC HYW1275835G
Ethyl trans-4-decenoate is an organic ester used as a biochemical reagent and research chemical. It is commonly handled as a liquid and applied in biochemical assays, fragrance and flavor research, and as a building block in organic synthesis.
- Used as a biochemical assay reagent and building block in organic synthesis.
- Molecular formula C12H22O2, molecular weight 198.30 g/mol.
- Typically a colorless clear liquid at room temperature.
- Soluble in alcohol, insoluble in water.
- Storage: stable at room temperature; in solution store at -80 °C for up to 2 years or -20 °C for up to 1 year.
- Purity commonly supplied at high assay levels; supplier listings report ranges from ≈96% to >98% depending on source.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.