Learn More
Medchemexpress LLC Ethylboronic acid | 4433-63-0 | 73.89 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW000933
Encompass_Preferred
Ethylboronic acid is an organic boron compound utilized as a drug intermediate. It is suitable for constructing macrocycles, capsules, polymers, and covalent organic frameworks (COFs).
  • Acts as a drug intermediate
  • Facilitates construction of macrocycles
  • Used in capsule synthesis
  • Enables polymer formation
  • Supports covalent organic framework (COF) creation
  • Applicable in biochemical assays
  • Key for self-assembly processes
  • Utilized in dynamic covalent chemistry
  • Forms supramolecular architectures
Catalog No. 50-004-50001