Learn More
Cayman Chemical EthylD-GlucuronIde-d5 5mg
SDP
Supplier:  Cayman Chemical 222735 MG
Encompass_Preferred
An internal standard for the quantification of ethyl-β-D-glucuronide by GC- or LC-MS
Catalog No. 50-296-4304