Learn More
Cayman Chemical Ethylen Glycol mondodecyl5mg
SDP
Supplier:  Cayman Chemical 368575 MG
Encompass_Preferred
A nonionic surfactant; CMC = 27.5 µM at 25°C; enhances the transdermal permeation of the antiemetic ondansetron across isolated porcine skin at 160 mM, however, it decreases the viability of human epidermal keratinocytes and human dermal fibroblasts at 160 µM
Catalog No. 50-290-9663